Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AZGP1
Entrez Gene ID
508800
Gene Name
alpha-2-glycoprotein 1, zinc-binding
Gene Aliases
ZAG
Chromosome Location
Chr.25: 36445777 - 36450266 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034331.1 NP_001029503.1 1 - 2 122 66
GenBank mRNA BC102237.1 - 1 - 2 122 66

View Full Product Details