Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RALA
Entrez Gene ID
538477
Gene Name
RAS like proto-oncogene A
Gene Aliases
-
Chromosome Location
Chr.4: 81185789 - 81244560 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034644.2 NP_001029816.1 4 - 5 711 59
GenBank mRNA BC102111.1 - 4 - 5 711 59
BT025381.1 - 4 - 5 763 59

View Full Product Details