Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SIGMAR1
Entrez Gene ID
538903
Gene Name
sigma non-opioid intracellular receptor 1
Gene Aliases
OPRS1
Chromosome Location
Chr.8: 76005565 - 76008280 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034655.1 NP_001029827.1 3 - 4 524 140
GenBank mRNA BT021620.1 - 3 - 4 524 140

View Full Product Details