Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TECR
Entrez Gene ID
614105
Gene Name
trans-2,3-enoyl-CoA reductase
Gene Aliases
GPSN2
Chromosome Location
Chr.7: 11232367 - 11259896 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034748.2 NP_001029920.1 5 - 6 355 77
GenBank mRNA BC102503.1 - 5 - 6 356 77

View Full Product Details