Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SKP1
Entrez Gene ID
615427
Gene Name
S-phase kinase associated protein 1
Gene Aliases
SKP1A
Chromosome Location
Chr.7: 45805398 - 45817464 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034781.2 NP_001029953.1 4 - 5 415 121
GenBank mRNA BC102435.1 - 4 - 5 418 121
BT029901.1 - 4 - 5 393 121

View Full Product Details