Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AIMP1
Entrez Gene ID
505126
Gene Name
aminoacyl tRNA synthetase complex interacting multifunctional protein 1
Gene Aliases
SCYE1, p43
Chromosome Location
Chr.6: 18837531 - 18864984 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035018.2 NP_001030190.2 4 - 5 469 126
GenBank mRNA BC103049.1 - 4 - 5 470 126

View Full Product Details