Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RBM8A
Entrez Gene ID
518045
Gene Name
RNA binding motif protein 8A
Gene Aliases
-
Chromosome Location
Chr.3: 21446207 - 21450620 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035324.2 NP_001030401.1 5 - 6 530 70
GenBank mRNA BC102484.1 - 5 - 6 531 70
BT025376.1 - 5 - 6 498 70

View Full Product Details