Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
FXYD7
Entrez Gene ID
616128
Gene Name
FXYD domain containing ion transport regulator 7
Gene Aliases
-
Chromosome Location
Chr.18: 45868662 - 45877788 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037627.1 NP_001032716.1 - 95 99
GenBank mRNA BC103263.1 - - 95 99

View Full Product Details