Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FAM78A
Entrez Gene ID
506709
Gene Name
family with sequence similarity 78 member A
Gene Aliases
-
Chromosome Location
Chr.11: 101450824 - 101465328 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038508.1 NP_001033597.1 2 - 3 1185 68
GenBank mRNA BT021704.1 - 2 - 3 1185 68

View Full Product Details