Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NAGA
Entrez Gene ID
533357
Gene Name
alpha-N-acetylgalactosaminidase
Gene Aliases
-
Chromosome Location
Chr.5: 113022843 - 113032870 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046349.1 NP_001039814.1 7 - 8 896 80
GenBank mRNA BC114813.1 - 7 - 8 922 80
BT021618.1 - 7 - 8 896 80

View Full Product Details