Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PPDPF
Entrez Gene ID
506990
Gene Name
pancreatic progenitor cell differentiation and proliferation factor
Gene Aliases
C13H20orf149, C20orf149
Chromosome Location
Chr.13: 54136186 - 54137875 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075294.1 NP_001068762.1 1 - 2 127 79
GenBank mRNA BC102578.1 - 1 - 2 127 79

View Full Product Details