Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TIMD4
Entrez Gene ID
507549
Gene Name
T cell immunoglobulin and mucin domain containing 4
Gene Aliases
-
Chromosome Location
Chr.7: 68286089 - 68314227 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075320.1 NP_001068788.1 1 - 2 75 108
GenBank mRNA BT020787.1 - 1 - 2 75 108

View Full Product Details