Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCNH
Entrez Gene ID
615512
Gene Name
cyclin H
Gene Aliases
-
Chromosome Location
Chr.7: 87037314 - 87067889 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079788.2 NP_001073256.1 3 - 4 436 119
GenBank mRNA BC103224.1 - 3 - 4 437 119

View Full Product Details