Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TDGF1
Entrez Gene ID
784029
Gene Name
teratocarcinoma-derived growth factor 1
Gene Aliases
-
Chromosome Location
Chr.22: 52867447 - 52872205 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080358.1 NP_001073827.1 5 - 6 652 75
GenBank mRNA BT021740.1 - 5 - 6 652 75

View Full Product Details