Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BCLAF3
Entrez Gene ID
511003
Gene Name
BCLAF1 and THRAP3 family member 3
Gene Aliases
CXHXorf23
Chromosome Location
Chr.X: 123192246 - 123244691 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BT021587.1 - 2 - 3 376 140

View Full Product Details