Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AMH
Entrez Gene ID
280718
Gene Name
anti-Mullerian hormone
Gene Aliases
-
Chromosome Location
Chr.7: 21401483 - 21404348 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173890.1 NP_776315.1 1 - 2 468 156

View Full Product Details