Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LDHB
Entrez Gene ID
281275
Gene Name
lactate dehydrogenase B
Gene Aliases
-
Chromosome Location
Chr.5: 88544523 - 88562770 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001316338.1 NP_001303267.1 6 - 7 857 112
NM_174100.2 NP_776525.2 6 - 7 857 112
GenBank mRNA AJ401268.1 - 6 - 7 821 112
BC102217.1 - 6 - 7 788 112
BC142006.1 - 6 - 7 858 112
BC151427.1 - 6 - 7 788 112
BT021009.1 - 6 - 7 825 112
EE338507.1 - 2 - 3 210 112

View Full Product Details