Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AIPL1
Entrez Gene ID
281609
Gene Name
aryl hydrocarbon receptor interacting protein like 1
Gene Aliases
-
Chromosome Location
Chr.19: 25422472 - 25431855 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174234.1 NP_776659.1 1 - 2 101 98
GenBank mRNA AF296410.1 - 1 - 2 101 98

View Full Product Details