Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CKMT1A
Entrez Gene ID
281692
Gene Name
creatine kinase, mitochondrial 1A
Gene Aliases
CKMT1, CKMT1B
Chromosome Location
Chr.21: 55322250 - 55328398 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174275.2 NP_776700.1 3 - 4 618 87
GenBank mRNA AB003307.1 - 3 - 4 618 87
BC102422.1 - 4 - 5 656 87

View Full Product Details