Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HINT3
Entrez Gene ID
281817
Gene Name
histidine triad nucleotide binding protein 3
Gene Aliases
-
Chromosome Location
Chr.9: 25324168 - 25340146 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174341.2 NP_776766.1 3 - 4 477 105
GenBank mRNA AF398236.1 - 3 - 4 477 105
BC110193.1 - 3 - 4 695 105

View Full Product Details