Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PRDX1
Entrez Gene ID
281997
Gene Name
peroxiredoxin 1
Gene Aliases
PRX1
Chromosome Location
Chr.3: 100551342 - 100565167 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174431.1 NP_776856.1 3 - 4 304 68
GenBank mRNA AB098940.1 - 3 - 4 297 68
AB099055.1 - 3 - 4 296 68
AF305561.1 - 3 - 4 304 68
BC148009.1 - 3 - 4 295 68
BT021073.1 - 3 - 4 292 68

View Full Product Details