Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VEGFC
Entrez Gene ID
282122
Gene Name
vascular endothelial growth factor C
Gene Aliases
-
Chromosome Location
Chr.27: 8017962 - 8103427 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174488.2 NP_776913.1 6 - 7 1266 127
GenBank mRNA AB004275.1 - 6 - 7 1266 127

View Full Product Details