Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CHRNA5
Entrez Gene ID
282177
Gene Name
cholinergic receptor nicotinic alpha 5 subunit
Gene Aliases
-
Chromosome Location
Chr.21: 30945440 - 30973322 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174514.2 NP_776939.1 1 - 2 298 100
GenBank mRNA AF487464.1 - 1 - 2 298 100

View Full Product Details