Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RASSF2
Entrez Gene ID
282337
Gene Name
Ras association domain family member 2
Gene Aliases
-
Chromosome Location
Chr.13: 47131128 - 47190109 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174593.2 NP_777018.1 9 - 10 920 89
GenBank mRNA AF277231.1 - 9 - 10 920 89

View Full Product Details