The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_174601.2 | NP_777026.1 | 2 - 3 | 1219 | 72 |
GenBank mRNA | AY039236.1 | - | 2 - 3 | 1219 | 72 |
BC123803.1 | - | 2 - 3 | 808 | 72 | |
LC005614.1 | - | 2 - 3 | 604 | 72 |