Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HSD11B2
Entrez Gene ID
282434
Gene Name
hydroxysteroid 11-beta dehydrogenase 2
Gene Aliases
-
Chromosome Location
Chr.18: 35036356 - 35046071 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174642.2 NP_777067.1 3 - 4 763 70
GenBank mRNA AF074706.2 - 3 - 4 765 70
BC102488.1 - 3 - 4 796 70

View Full Product Details