Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CYHR1
Entrez Gene ID
282612
Gene Name
cysteine and histidine rich 1
Gene Aliases
-
Chromosome Location
Chr.14: 478700 - 492655 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001322822.1 NP_001309751.1 4 - 5 1312 98

View Full Product Details