Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CHIA
Entrez Gene ID
282645
Gene Name
chitinase acidic
Gene Aliases
-
Chromosome Location
Chr.3: 32071071 - 32091157 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174699.2 NP_777124.1 10 - 11 1190 105
GenBank mRNA AB051629.1 - 10 - 11 1190 105
BC102931.1 - 10 - 11 1190 105

View Full Product Details