Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NDUFA11
Entrez Gene ID
326346
Gene Name
NADH:ubiquinone oxidoreductase subunit A11
Gene Aliases
b14.7
Chromosome Location
Chr.7: 18474080 - 18479129 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175718.2 NP_783649.1 3 - 4 367 81
GenBank mRNA AJ510148.1 - 3 - 4 367 81
BC102976.1 - 3 - 4 388 81

View Full Product Details