Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RHOA
Entrez Gene ID
338049
Gene Name
ras homolog family member A
Gene Aliases
ARHA
Chromosome Location
Chr.22: 50701226 - 50751133 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_176645.3 NP_788818.1 3 - 4 395 77
GenBank mRNA BC102880.1 - 3 - 4 395 77
M27278.1 - 3 - 4 359 77

View Full Product Details