Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GGTA1
Entrez Gene ID
281780
Gene Name
alpha-galactosyltransferase 1 (glycoprotein)
Gene Aliases
-
Chromosome Location
Chr.11: 92306752 - 92395084 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177511.2 NP_803477.1 7 - 8 744 82
GenBank mRNA BC151691.1 - 6 - 7 456 82
J04989.1 - 7 - 8 744 82

View Full Product Details