Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RPS18
Entrez Gene ID
326602
Gene Name
ribosomal protein S18
Gene Aliases
-
Chromosome Location
Chr.23: 7446290 - 7451048 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033614.2 NP_001028786.1 3 - 4 232 82
GenBank mRNA AB098889.1 - 3 - 4 213 82
AB098899.1 - 2 - 3 173 82
AB099024.1 - 3 - 4 216 82
BC102293.1 - 3 - 4 233 82

View Full Product Details