Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RPS18
Entrez Gene ID
326602
Gene Name
ribosomal protein S18
Gene Aliases
-
Chromosome Location
Chr.23: 7446290 - 7451048 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033614.2 NP_001028786.1 5 - 6 422 89
GenBank mRNA AB098889.1 - 5 - 6 403 89
AB098899.1 - 4 - 5 363 89
AB099024.1 - 5 - 6 406 89
BC102293.1 - 5 - 6 423 89

View Full Product Details