Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ADIPOR1
Entrez Gene ID
407235
Gene Name
adiponectin receptor 1
Gene Aliases
-
Chromosome Location
Chr.16: 79870209 - 79883997 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034055.1 NP_001029227.1 3 - 4 440 69
GenBank mRNA BC102259.1 - 3 - 4 440 69

View Full Product Details