Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
POLR2C
Entrez Gene ID
504452
Gene Name
RNA polymerase II subunit C
Gene Aliases
-
Chromosome Location
Chr.18: 25458202 - 25466511 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034212.2 NP_001029384.1 6 - 7 466 63
GenBank mRNA BC102301.1 - 6 - 7 468 63
BT030680.1 - 6 - 7 462 63

View Full Product Details