Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GATA5
Entrez Gene ID
504784
Gene Name
GATA binding protein 5
Gene Aliases
-
Chromosome Location
Chr.13: 54799159 - 54809119 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034221.2 NP_001029393.1 2 - 3 812 94
GenBank mRNA BC102821.1 - 2 - 3 813 94

View Full Product Details