Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TSEN34
Entrez Gene ID
505571
Gene Name
tRNA splicing endonuclease subunit 34
Gene Aliases
-
Chromosome Location
Chr.18: 63209093 - 63213101 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034251.1 NP_001029423.1 1 - 2 181 78
GenBank mRNA BC105271.1 - 1 - 2 181 78

View Full Product Details