Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TSEN34
Entrez Gene ID
505571
Gene Name
tRNA splicing endonuclease subunit 34
Gene Aliases
-
Chromosome Location
Chr.18: 63209093 - 63213101 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034251.1 NP_001029423.1 3 - 4 685 135
GenBank mRNA BC105271.1 - 3 - 4 685 135

View Full Product Details