Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CNPY4
Entrez Gene ID
505637
Gene Name
canopy FGF signaling regulator 4
Gene Aliases
CNPY
Chromosome Location
Chr.25: 36333373 - 36337700 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034253.2 NP_001029425.1 4 - 5 562 68
GenBank mRNA BC104626.1 - 4 - 5 566 68

View Full Product Details