The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001034253.2 | NP_001029425.1 | 5 - 6 | 675 | 67 |
GenBank mRNA | BC104626.1 | - | 5 - 6 | 679 | 67 |