Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OCIAD2
Entrez Gene ID
505877
Gene Name
OCIA domain containing 2
Gene Aliases
-
Chromosome Location
Chr.6: 67528017 - 67539933 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034258.2 NP_001029430.1 4 - 5 292 113
GenBank mRNA BC103326.1 - 4 - 5 295 113

View Full Product Details