Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ECSIT
Entrez Gene ID
507245
Gene Name
ECSIT signaling integrator
Gene Aliases
-
Chromosome Location
Chr.7: 15857760 - 15874392 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034286.1 NP_001029458.1 3 - 4 592 83
GenBank mRNA BC104572.1 - 3 - 4 592 83

View Full Product Details