Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EFNA1
Entrez Gene ID
507319
Gene Name
ephrin A1
Gene Aliases
-
Chromosome Location
Chr.3: 15466565 - 15473164 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034292.1 NP_001029464.1 2 - 3 492 74
GenBank mRNA BC102889.2 - 2 - 3 492 74

View Full Product Details