Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NIPA2
Entrez Gene ID
507430
Gene Name
NIPA magnesium transporter 2
Gene Aliases
-
Chromosome Location
Chr.2: 1044499 - 1066244 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034298.1 NP_001029470.1 2 - 3 58 71
GenBank mRNA BC104627.1 - 2 - 3 58 71

View Full Product Details