The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001034328.2 | NP_001029500.1 | 3 - 4 | 1260 | 88 |
GenBank mRNA | BC102143.1 | - | 3 - 4 | 1261 | 88 |