Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SAT1
Entrez Gene ID
508861
Gene Name
spermidine/spermine N1-acetyltransferase 1
Gene Aliases
SAT
Chromosome Location
Chr.X: 119431931 - 119434979 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034333.2 NP_001029505.1 4 - 5 484 96
GenBank mRNA BC102304.1 - 4 - 5 485 96

View Full Product Details