Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ERH
Entrez Gene ID
508901
Gene Name
ERH mRNA splicing and mitosis factor
Gene Aliases
-
Chromosome Location
Chr.10: 81172324 - 81185012 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034336.1 NP_001029508.1 1 - 1 254 116
GenBank mRNA BC102967.1 - 1 - 1 254 116

View Full Product Details