Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IL11RA
Entrez Gene ID
508932
Gene Name
interleukin 11 receptor subunit alpha
Gene Aliases
-
Chromosome Location
Chr.8: 76017709 - 76027359 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034339.1 NP_001029511.1 6 - 7 652 80
GenBank mRNA BC103219.1 - 6 - 7 652 80

View Full Product Details