Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ETHE1
Entrez Gene ID
509150
Gene Name
ETHE1 persulfide dioxygenase
Gene Aliases
-
Chromosome Location
Chr.18: 51712253 - 51729580 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034344.1 NP_001029516.1 4 - 5 552 109
GenBank mRNA BC102496.1 - 4 - 5 552 109

View Full Product Details