Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NIFK
Entrez Gene ID
509598
Gene Name
nucleolar protein interacting with the FHA domain of MKI67
Gene Aliases
MKI67IP
Chromosome Location
Chr.2: 73387424 - 73397871 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034354.2 NP_001029526.1 2 - 3 280 94
GenBank mRNA BC102791.1 - 2 - 3 283 94

View Full Product Details